Reagents Potassium Hydroxide, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Potassium Hydroxide, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2285000-500A5| Item: UX-78913-09

Manufactured using the highest quality raw materials in ISO 9001 facility

$70.90USD / Each

$70.90USD / Each
Product Description

Key Features

  • White deliquescent solid
  • Density: 2.04
  • Boiling point: approximately 1320 °C (2408 °F)
  • Solubility information: 1210 g/L at 25 °C (77 °F)
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaKOH
  • ClassBase
  • FormSolid
  • Weight (g)500
  • Formula Weight56.11
  • PackagingWide-mouth container
  • DOT InformationPOTASSIUM HYDROXIDE, SOLID, 8, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1813
  • CAS Number1310-58-3
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPotassium Hydroxide, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A