Reagents® Potassium Hydroxide, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • White deliquescent solid
  • Density: 2.04
  • Boiling point: approximately 1320 °C (2408 °F)
  • Solubility information: 1210 g/L at 25 °C (77 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$70.90 - $357.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Potassium Hydroxide, Purified, Natural Poly Container, 3 kg EW-62285-47 Mfr # C2285000-3A5
COMPARE
Base Poly container 3000
$357.50USD / Each
Reagents Potassium Hydroxide, Purified, Wide-Mouth Container, 500 g EW-78913-09 Mfr # C2285000-500A5
COMPARE
Base Wide-mouth container 500
$70.90USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat