Reagents Potassium Iodate, ACS Reagent, Wide-Mouth Container, 500 g

Representative image only.

Reagents Potassium Iodate, ACS Reagent, Wide-Mouth Container, 500 g

Reagents | Mfr: C1285500-500A5| Item: EW-62282-50

Manufactured using the highest quality raw materials in ISO 9001 facility

$700.00USD / Each

$700.00USD / Each
Product Description

Key Features

  • Minimum Assay 99.4%
  • Colorless/white solid
  • Density: 3.89
  • Melting point: 560 °C (1040 °F), decomposes
  • Solubility information: 81 g/L at 20 °C (68 °F)
  • pH range: 5 to 8
  • 500 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKIO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)500
  • Formula Weight214
  • PackagingWide-mouth container
  • DOT InformationOXIDIZING SOLID, N.O.S. (POTASSIUM IODATE), 5.1, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1479
  • CAS Number7758-05-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionPotassium Iodate, ACS Reagent, Wide-Mouth Container, 500 g
Documents & Certificates
Reviews
Q&A