Reagents Potassium Nitrate, ACS Reagent, Natural Poly Container, 250 g

Representative image only.

Reagents Potassium Nitrate, ACS Reagent, Natural Poly Container, 250 g

Reagents | Mfr: C1286500-250A6| Item: EW-62282-59

Manufactured using the highest quality raw materials in ISO 9001 facility

$104.25USD / Each

$104.25USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • White crystals
  • Density: 2.11
  • Boiling point: approximately 400 °C (752 °F)
  • Solubility information: 383 g/L at 25 °C (77 °F)
  • pH range: 4.5 to 8.5
  • 250 g natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKNO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)250
  • Formula Weight101.1
  • PackagingNatural poly container
  • DOT InformationPOTASSIUM NITRATE, 5.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1486
  • CAS Number7757-79-1
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)334
  • Melting Point F (° F)633.2
  • DescriptionPotassium Nitrate, ACS Reagent, Natural Poly Container, 250 g
Documents & Certificates
Reviews
Q&A