Reagents Potassium Permanganate, 0.20N, 1L, Standardized

Representative image only.

Reagents Potassium Permanganate, 0.20N, 1L, Standardized

Reagents | Mfr: CP208500-1C| Item: EW-78918-12

Manufactured using the highest quality raw materials in ISO 9001 facility

$52.40USD / Each

$52.40USD / Each
Product Description

Key Features

  • N/5, Assay 0.1995 to 0.2005 Normal, Aqueous
  • 18 month warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

18 Month Warranty
Specifications
  • Normality0.2
  • ClassStandard
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)1000
  • PackagingAmber Glass Bottle
  • Shelf Life (days)555
  • Shelf Life (months)18
  • DescriptionPotassium Permanganate, 0.20N, 1L, Standardized
  • Warranty18 months
Documents & Certificates
Reviews
Q&A