Reagents Potassium Permanganate, Purified, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Potassium Permanganate, Purified, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2288500-2.5A5| Item: EW-78919-25

Manufactured using the highest quality raw materials in ISO 9001 facility

$208.50USD / Each

$208.50USD / Each
Product Description

Key Features

  • Purple-black solid
  • Density: 2.7
  • Melting point: >240 °C (>464 °F)
  • Solubility information: 76 g/L at 25 °C (77 °F)
  • 2.5 kg natural poly container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaKMnO4
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight158.03
  • PackagingPoly container
  • DOT InformationPOTASSIUM PERMANGANATE, 5.1, P.G. II
  • Storage SymbolAmbient
  • CAS Number7722-64-7
  • UN NumberUN1490
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPotassium Permanganate, Purified, Natural Poly Container, 2.5 kg
  • Warranty10 year
Documents & Certificates
Reviews
Q&A