Reagents Potassium Sodium Tartrate, ACS Reagent, Natural Poly Bottle, 3 kg

Representative image only.

Reagents Potassium Sodium Tartrate, ACS Reagent, Natural Poly Bottle, 3 kg

Reagents | Mfr: C1290500-3A5| Item: EW-62282-71

Manufactured using the highest quality raw materials in ISO 9001 facility

$522.00USD / Each

$522.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white powder
  • Density: 1.79
  • Boiling point: 220 °C (428 °F), decomposes
  • Melting point: 70 to 80 °C (158 to 176 °F)
  • 3 kg natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKNaC4H4O6·4H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsRochelle salt
  • Weight (g)3000
  • Formula Weight282.22
  • PackagingPoly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6381-59-5
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionPotassium Sodium Tartrate, ACS Reagent, Natural Poly Bottle, 3 kg
Documents & Certificates
Reviews
Q&A