Reagents Potassium Sulfate, ACS Reagent, Container, 2.5 kg

Representative image only.

Reagents Potassium Sulfate, ACS Reagent, Container, 2.5 kg

Reagents | Mfr: C1291000-2.5KG| Item: EW-78920-46

Manufactured using the highest quality raw materials in ISO 9001 facility

$456.00USD / Each

$456.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white powder
  • Density: 2.66
  • Solubility information: soluble
  • 2.5 kg container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaK2SO4
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight174.26
  • PackagingContainer
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7778-80-5
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)1689
  • Boiling Point F (° F)3072.2
  • Melting Point C (° C)1069
  • Melting Point F (° F)1956.2
  • DescriptionPotassium Sulfate, ACS Reagent, Container, 2.5 kg
  • Warranty5 years
Documents & Certificates
Reviews
Q&A