Reagents Potassium Sulfate, Low Nitrogen, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Potassium Sulfate, Low Nitrogen, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2291000-500A5| Item: EW-78913-34

Manufactured using the highest quality raw materials in ISO 9001 facility

$132.50USD / Each

$132.50USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.66
  • Solubility information: soluble
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaK2SO4
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight174.26
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7778-80-5
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)1069
  • Melting Point F (° F)1956.2
  • DescriptionPotassium Sulfate, Low Nitrogen, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A