Reagents Sodium Acetate Anhydrous, ACS Reagent, Fiber Drum, 12 kg

Representative image only.

Reagents Sodium Acetate Anhydrous, ACS Reagent, Fiber Drum, 12 kg

Reagents | Mfr: C1306400-12F4| Item: EW-62282-77

Manufactured using the highest quality raw materials in ISO 9001 facility

$1,150.00USD / Each

$1,150.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white powder
  • Density: 1.53
  • Solubility information: 465 g/L at 20 °C (68 °F)
  • pH range: 7 to 9.2
  • 12 kg fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNaC2H3O2
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)12000
  • Formula Weight82.03
  • PackagingFiber drum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number127-09-3
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)123
  • Boiling Point F (° F)253.4
  • Melting Point C (° C)324
  • Melting Point F (° F)615.2
  • DescriptionSodium Acetate Anhydrous, ACS Reagent, Fiber Drum, 12 kg
Documents & Certificates
Reviews
Q&A