Reagents Sodium Acetate Trihydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Sodium Acetate Trihydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C1306500-100C2| Item: EW-78913-53

Manufactured using the highest quality raw materials in ISO 9001 facility

$40.40USD / Each

$40.40USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white crystal
  • Density: 1.45
  • Solubility information: 762 g/L at 20 °C (68 °F)
  • pH range: 7.5 to 9.2
  • 100 g wide-mouth amber glass bottle
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNaC2H3O2·3H2O
  • ClassSalts
  • SynonymsSodium acetate, 3-hydrate
  • FormSolid
  • Weight (g)100
  • Formula Weight136.08
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6131-90-4
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)58
  • Melting Point F (° F)136.4
  • DescriptionSodium Acetate Trihydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A