Reagents Sodium Bicarbonate, Purified, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Sodium Bicarbonate, Purified, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2308500-2.5A5| Item: EW-78919-31

Manufactured using the highest quality raw materials in ISO 9001 facility

$58.40USD / Each

$58.40USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.16
  • Melting point: 270 °C (518 °F) with decomposition
  • Solubility information: 103 g/L at 25 °C (77 °F)
  • 2.5 kg natural poly container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNaHCO3
  • ClassSalts
  • FormSolid
  • SynonymsBicarbonate of soda, Sodium acid carbonate, Monosodium carbonate, Sodium hydrogen carbonate, Carbonic acid monosodium salt, Baking soda
  • Weight (g)2500
  • Formula Weight84.01
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number144-55-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSodium Bicarbonate, Purified, Natural Poly Container, 2.5 kg
  • Warranty10 year
Documents & Certificates
Reviews
Q&A