Reagents Sodium Chromate Anhydrous, Purified, Natural Poly Container, 3 kg

Representative image only.

Reagents Sodium Chromate Anhydrous, Purified, Natural Poly Container, 3 kg

Reagents | Mfr: C2314000-3A5| Item: EW-62285-72

Manufactured using the highest quality raw materials in ISO 9001 facility

$695.00USD / Each

$695.00USD / Each
Product Description

Key Features

  • Yellow solid
  • Density: 2.72
  • Solubility information: 876 g/L at 25 °C (77 °F)
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2CrO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)3000
  • Formula Weight161.9732
  • PackagingPoly container
  • DOT InformationOXIDIZING SOLID, CORROSIVE, N.O.S. (SODIUM CHROMATE), 5.1 (8), P.G. III
  • Storage SymbolAmbient
  • CAS Number7775-11-3
  • UN NumberUN3085
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)792
  • Melting Point F (° F)1457.6
  • DescriptionSodium Chromate Anhydrous, Purified, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A