Reagents Sodium Dithionite, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Sodium Dithionite, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C2316300-500C2| Item: EW-78919-04

Manufactured using the highest quality raw materials in ISO 9001 facility

$68.60USD / Each

$68.60USD / Each
Product Description

Key Features

  • White to off-white powder
  • Density: 0.88
  • Boiling point: >80 °C (176 °F), decomposes
  • Melting point: 52 °C (125.6 °F), decomposes
  • Flash point: >100 °C (>212 °F)
  • Solubility information: 241 g/L at 20 °C (68 °F)
  • 500 g wide-mouth amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNa2O4S2
  • ClassSalts
  • SynonymsSodium hydrosulfite
  • FormSolid
  • Weight (g)500
  • Formula Weight174.11
  • PackagingWide-mouth amber glass bottle
  • DOT InformationSODIUM DITHIONITE, 4.2, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1384
  • CAS Number7775-14-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Dithionite, Purified, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty5 years
Documents & Certificates
Reviews
Q&A