Reagents Sodium Hydroxide, 50% (w/w), Reagent, Natural Poly Container, 4 L

Representative image only.

Reagents Sodium Hydroxide, 50% (w/w), Reagent, Natural Poly Container, 4 L

Reagents | Mfr: C1318600-4A| Item: EW-78925-41

Manufactured using the highest quality raw materials in ISO 9001 facility

$194.00USD / Each

$194.00USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 1.53
  • Solubility information: miscible
  • pH range: 14
  • 4 L natural poly container
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

3 Year Warranty
Specifications
  • Purity GradeReagent
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • Vapor Pressure (torr)13
  • PackagingNatural poly container
  • DOT InformationSODIUM HYDROXIDE SOLUTION, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number1310-73-2
  • UN NumberUN1824
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)140
  • Boiling Point F (° F)284
  • Melting Point C (° C)12
  • Melting Point F (° F)53.6
  • DescriptionSodium Hydroxide, 50% (w/w), Reagent, Natural Poly Container, 4 L
  • Warranty3 year
Documents & Certificates
Reviews
Q&A