Reagents Sodium Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Sodium Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C1320500-100C2| Item: EW-78913-95

Manufactured using the highest quality raw materials in ISO 9001 facility

$59.00USD / Each

$59.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white crystals
  • Density: 2.26
  • Boiling point: 380 °C (716 °F) with decomposition
  • Solubility information: 874 g/L at 20 °C (68 °F)
  • pH range: 5.5 to 8.3
  • 100 g wide-mouth amber glass bottle
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNaNO3
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight84.99
  • PackagingWide-mouth amber glass bottle
  • DOT InformationSODIUM NITRATE, 5.1, P.G. III
  • UN NumberUN1498
  • CAS Number7631-99-4
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)306
  • Melting Point F (° F)582.8
  • DescriptionSodium Nitrate, 100g, ACS Reagent
  • Warranty7 year
Documents & Certificates
Reviews
Q&A