Reagents Sodium Nitrite, 2.5kg, ACS Reagent

Representative image only.

Reagents Sodium Nitrite, 2.5kg, ACS Reagent

Reagents | Mfr: C1321000-2.5A5| Item: UX-78919-42

Manufactured using the highest quality raw materials in ISO 9001 facility

$429.75USD / Each

$429.75USD / Each

Buy It Together

Current Item

Reagents Sodium Nitrite, 2.5kg, ACS Reagent
Reagents Sodium Nitrite, 2.5kg, ACS Reagent
7891942

$429.75USD / Each
+
Cole-Parmer Sodium Nitrite, ACS Grade, 500 g
8840485

$80.20USD / Each
+ =
Total Price 572.95 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Minimum Assay 97%, Crystal
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaNaNO2
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight69
  • PackagingPoly Jar
  • DOT InformationSODIUM NITRITE, 5.1 (6.1)
  • CAS Number7632-00-0
  • UN NumberUN1500
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Nitrite, 2.5kg, ACS Reagent
  • Warranty5 years
Documents & Certificates
Reviews
Q&A