Reagents Sodium Sulfate Anhydrous, Low Organic, ACS Reagent, Fiber Drum, 50 kg

Representative image only.

Reagents Sodium Sulfate Anhydrous, Low Organic, ACS Reagent, Fiber Drum, 50 kg

Reagents | Mfr: C1325550-50F4| Item: EW-62283-14

Manufactured using the highest quality raw materials in ISO 9001 facility

$3,640.00USD / Each

$3,640.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Low organic
  • White solid
  • Density: 2.7
  • Boiling point: >890 °C (>1634 °F) with decomposition
  • Melting point: 884 to 888 °C (1623.2 to 1630.4 °F)
  • Solubility information: 200 g/L at 20 °C (68 °F)
  • pH range: 5.2 to 9.2
  • Low organic
  • 50 kg fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNa2SO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)50000
  • Formula Weight142.04
  • PackagingFiber drum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7757-82-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Sulfate Anhydrous, Low Organic, ACS Reagent, Fiber Drum, 50 kg
Documents & Certificates
Reviews
Q&A