Reagents Stannous Chloride Dihydrate, ACS Reagent, Wide-Mouth Container, 500 g

Representative image only.

Reagents Stannous Chloride Dihydrate, ACS Reagent, Wide-Mouth Container, 500 g

Reagents | Mfr: C1330000-500A5| Item: EW-78914-22

Manufactured using the highest quality raw materials in ISO 9001 facility

$281.50USD / Each

$281.50USD / Each
Product Description

Key Features

  • White to off-white solid
  • Density: 2.71
  • Melting point: 37 to 38 °C (98.6 to 100.4 °F), decomposes
  • Solubility information: 1780 g/L at 10 °C (50 °F)
  • 500 g wide-mouth container
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Two year warranty
Specifications
  • Purity GradeACS
  • Linear FormulaSnCl2·2H2O
  • ClassSalts
  • FormSolid
  • SynonymsTin (II) chloride 2-hydrate
  • Weight (g)500
  • Formula Weight225.65
  • PackagingWide-mouth container
  • DOT InformationCORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. (TIN CHLORIDE), 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number10025-69-1
  • UN NumberUN3260
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)652
  • Boiling Point F (° F)1205.6
  • DescriptionStannous Chloride Dihydrate, ACS Reagent, Wide-Mouth Container, 500 g
  • Warranty2 year
Documents & Certificates
Reviews
Q&A