Reagents Sulfuric Acid, 70% (w/w), Textile Grade, Natural Poly Bottle, 4 L

Representative image only.

Reagents Sulfuric Acid, 70% (w/w), Textile Grade, Natural Poly Bottle, 4 L

Reagents | Mfr: C7306600-4A| Item: EW-78925-42

Manufactured using the highest quality raw materials in ISO 9001 facility

$110.25USD / Each

$110.25USD / Each
Product Description

Key Features

  • Colorless to tan liquid
  • Density: 1.61
  • Solubility information: miscible
  • pH range: <0.1
  • 4 L natural poly bottle
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradeTextile
  • Concentration70% w/w
  • Linear FormulaH2SO4
  • ClassVolumetric
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly bottle
  • DOT InformationSULFURIC ACID SOLUTION, 8, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN2796
  • CAS Number7664-93-9
  • Shelf Life (days)730
  • Shelf Life (months)24
  • Boiling Point C (° C)165
  • Boiling Point F (° F)329
  • DescriptionSulfuric Acid, 70% (w/w), Textile Grade, Natural Poly Bottle, 4 L
  • Warranty10 year
Documents & Certificates
Reviews
Q&A