Reagents Zobell's Solution, 4L

Representative image only.

Reagents Zobell's Solution, 4L

Reagents | Mfr: CZ000100-4B| Item: EW-78922-53

Manufactured using the highest quality raw materials in ISO 9001 facility

$379.50USD / Each

$379.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • ClassMixture
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly Bottle
  • Shelf Life (days)90
  • Shelf Life (months)2
  • DescriptionZobell's Solution, 4L,
Documents & Certificates
Reviews
Q&A