Retsch 01.462.0232 Mixer Jar Mill, Agate; 5 mL

Representative image only.

Retsch 01.462.0232 Mixer Jar Mill, Agate; 5 mL

Retsch | Mfr: 01.462.0232| Item: EW-04182-37

$4,535.00USD / Each

$4,535.00USD / Each

Buy It Together

Current Item

Retsch 01.462.0232 Mixer Jar Mill, Agate; 5 mL
Retsch 01.462.0232 Mixer Jar Mill, Agate; 5 mL
0418237

$4,535.00USD / Each
+ =
Total Price 4538.70 ADD 2 ITEMS TO CART
Product Description

More About this Item

Jars and media to meet your application needs.

Specifications
  • Sample Capacity (mL)5
  • MaterialAgate
  • DescriptionMixer Jar Mill, Agate; 5 mL
Reviews
Q&A