Retsch 02.462.0213 25 mL Mill Jar; stainless steel

Representative image only.

Retsch 02.462.0213 25 mL Mill Jar; stainless steel

Retsch | Mfr: 14620213| Item: EW-04182-32

$1,680.00USD / Each

$1,680.00USD / Each

Buy It Together

Current Item

Retsch 02.462.0213 25 mL Mill Jar; stainless steel
Retsch 02.462.0213 25 mL Mill Jar; stainless steel
0418232

$1,680.00USD / Each
+ =
Total Price 1683.70 ADD 2 ITEMS TO CART
Product Description

More About this Item

Jars and media to meet your application needs.

Specifications
  • Sample Capacity (mL)25
  • MaterialStainless Steel
  • Description25 mL Mill Jar; stainless steel
Reviews
Q&A