Ricca Chemical Company 1785-16 Calcium Hydroxide TS, Saturated Aqueous Solution (500 mL)

Representative image only.

Ricca Chemical Company 1785-16 Calcium Hydroxide TS, Saturated Aqueous Solution (500 mL)

Ricca Chemical Company | Mfr: 1785-16| Item: EW-86970-76

Quality and consistency your lab can count on

$96.10USD / Each

$96.10USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • CALCIUM HYDROXIDE TS

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • Packaging500 mL Poly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 1305-62-0 and 7732-18-5
  • Shelf Life (months)12
  • DescriptionCalcium Hydroxide TS, Saturated Aqueous Solution (500 mL)
Reviews
Q&A