Ricca Chemical Company 3580-1 Hydrochloric Acid, 50% (v/v) Aqueous Solution (4 L)

Representative image only.

Ricca Chemical Company 3580-1 Hydrochloric Acid, 50% (v/v) Aqueous Solution (4 L)

Ricca Chemical Company | Mfr: 3580-1| Item: EW-86986-03

High quality, competitive pricing, and rapid service

$186.10USD / Each

$186.10USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • HYDROCHLORIC ACID, 50% (v/v)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeComponents all ACS
  • Concentration0.5
  • Empirical FormulaHCl
  • ClassGeneral Solutions
  • FormLiquid
  • SynonymsMuriatic acid
  • Volume (mL)4000
  • Formula Weight36.45
  • PackagingPoly Bottle
  • DOT InformationHydrochloric Acid Solution; Class 8, Packing Group II
  • CAS NumberIncludes 7647-01-0 and 7732-18-5
  • UN NumberUN1789
  • Shelf Life (months)18
  • DescriptionHydrochloric Acid, 50% (v/v) Aqueous Solution (1 + 1); (4 L)
Reviews
Q&A