Ricca Chemical Company 5464.6-4 ORP Standard, +600 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution); 120 mL

Representative image only.

Ricca Chemical Company 5464.6-4 ORP Standard, +600 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution); 120 mL

Ricca Chemical Company | Mfr: 5464.6-4| Item: EW-86991-60

Get accurate measurements every time

$72.70USD / Each

$72.70USD / Each
Product Description

Key Features

  • Ready-made standard
  • ORP STD., 600 mV

More About this Item

The solution provides precise control allowing you to produce accurate measurements every time.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeComponents all ACS
  • ClassStandards
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)120
  • PackagingAmber Glass
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7783-83-7, 7783-85-9, 7664-93-9 and 7732-18-5
  • Shelf Life (months)9
  • DescriptionORP Standard, +600 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (120 mL)
Reviews
Q&A