Ricca Chemical Company 7200-16 Sodium Chloride, 0.85% (w/v) Aqueous Solution, Normal Saline (500 mL)

Representative image only.

Ricca Chemical Company 7200-16 Sodium Chloride, 0.85% (w/v) Aqueous Solution, Normal Saline (500 mL)

Ricca Chemical Company | Mfr: 7200-16| Item: EW-86975-81

Quality and consistency your lab can count on

$30.90USD / Each

$30.90USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SODIUM CHLORIDE, 0.85%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7647-14-5 and 7732-18-5
  • Shelf Life (months)24
  • DescriptionSodium Chloride, 0.85% (w/v) Aqueous Solution, Normal Saline (500 mL)
Reviews
Q&A