Ricca Chemical Company 7472-32 Sodium Hydroxide, 0.100 Normal (N/10) in Methanol (32 oz)

Representative image only.

Ricca Chemical Company 7472-32 Sodium Hydroxide, 0.100 Normal (N/10) in Methanol (32 oz)

Ricca Chemical Company | Mfr: 7472-32| Item: EW-86983-06

High quality, competitive pricing, and rapid service

$123.50USD / Each

$123.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SOD. HYDROXIDE, 0.1 N/MEOH

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality0.1
  • Purity GradeComponents all ACS
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • SynonymsCaustic Soda Solution
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationFlammable Liquid, n.o.s., (Methanol); Class 3, Packing Group II
  • UN NumberUN1993
  • CAS NumberIncludes 67-56-1 and 1310-73-2
  • Shelf Life (months)12
  • DescriptionSodium Hydroxide, 0.100 Normal (N/10) in Methanol (32 oz)
Reviews
Q&A