Ricca Chemical Company 8363-5 Synthetic Seawater, ASTM D 1141 Substitute Ocean Water, without Heavy Metals (20 L)

Representative image only.

Ricca Chemical Company 8363-5 Synthetic Seawater, ASTM D 1141 Substitute Ocean Water, without Heavy Metals (20 L)

Ricca Chemical Company | Mfr: 8363-5| Item: EW-86994-70

Quality and consistency your lab can count on

$352.90USD / Each

$352.90USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SYNTHETIC SEAWATER
  • ASTM D1141

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 10043-35-3, 10035-04-8, 7791-18-6, 7758-02-3, 7447-40-7, 144-55-8, 7647-14-5, 7681-49-4, 1310-73-2, 7757-82-6, 10025-70-4, and 7732-18-5
  • Shelf Life (months)12
  • DescriptionSynthetic Seawater, ASTM D 1141 Substitute Ocean Water, without Heavy Metals (20 L)
Reviews
Q&A