Ricca Chemical Company Hydrochloric Acid, 20% v/v 20 L

Representative image only.

Ricca Chemical Company Hydrochloric Acid, 20% v/v 20 L

Ricca Chemical Company | Mfr: R3574000-20F| Item: EW-20871-52

High quality, competitive pricing, and rapid service

$270.30USD / Each

$270.30USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.2
  • FormLiquid
  • Volume (mL)20000
  • Shelf Life (days)1080
  • Shelf Life (months)36
  • DescriptionHydrochloric Acid, 20% v/v 20 L
Documents & Certificates
Reviews
Q&A