Ricca Chemical Company Sodium Hydroxide, 0.3125 Normal, Poly Container; 4 L

Representative image only.

Ricca Chemical Company Sodium Hydroxide, 0.3125 Normal, Poly Container; 4 L

Ricca Chemical Company | Mfr: 7374-1| Item: EW-86987-79

High quality, competitive pricing, and rapid service

$101.00USD / Each

$101.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • SODIUM HYDROXIDE, 0.3125 N

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality0.3125
  • Purity GradeComponents all ACS
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • SynonymsCaustic Soda Solution
  • Volume (mL)4000
  • PackagingPoly Bottle
  • DOT InformationSodium Hydroxide Solution; Class 8, Packing Group III
  • CAS NumberIncludes 1310-73-2 and 7732-18-5
  • UN NumberUN1824
  • Shelf Life (months)24
  • DescriptionSodium Hydroxide, 0.3125 Normal, 4L, poly container
Reviews
Q&A