Ricca Chemical Company VeriSpec™ Phosphorus Standard for ICP 1000 ppm in H₂O; 100 mL

Representative image only.

Ricca Chemical Company VeriSpec™ Phosphorus Standard for ICP 1000 ppm in H₂O; 100 mL

Ricca Chemical Company | Mfr: RV010300-100N| Item: EW-85004-39

Single-element calibration standard

$126.30USD / Each

$126.30USD / Each
Product Description

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

More About this Item

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

For sale in the US only

Specifications
  • Concentration1000 ppm
  • Purity GradeISO 17025 and ISO Guide 34
  • ClassSpectroscopy
  • MediumWater
  • ChemistryStandards
  • Element/IonPhosphorus
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Shelf Life (days)720
  • Shelf Life (months)24
  • DescriptionPhosphorus Standard for ICP 1000 ppm in H₂O; 100 mL
Reviews
Q&A