Ricca Chemical Company VeriSpec™ Titanium Standard for ICP 1000 ppm 5% HCl/HF; 250 mL

Representative image only.

Ricca Chemical Company VeriSpec™ Titanium Standard for ICP 1000 ppm 5% HCl/HF; 250 mL

Ricca Chemical Company | Mfr: RV010322-250N| Item: EW-85005-24

Single-element calibration standard

$224.70USD / Each

$224.70USD / Each
Product Description

Key Features

  • Made to Guide 34 Standards!
  • Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

More About this Item

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

For sale in the US only

Specifications
  • Concentration1000 ppm
  • Purity GradeISO 17025 and ISO Guide 34
  • ClassSpectroscopy
  • ChemistryStandards
  • Medium5% Hydrochloric Acid and Hydrofluoric Acid
  • Element/IonTitanium
  • Volume (mL)250
  • PackagingPoly Bottle
  • DOT InformationPacking Group 2, Packing Class 8, Corrosive liquid acidic inorganic n.o.s. HCl Acid HF Acid Titanium III Chloride
  • UN Number3264
  • Shelf Life (days)720
  • Shelf Life (months)24
  • DescriptionTitanium Standard for ICP 1000 ppm 5% HCl/HF; 250 mL
Reviews
Q&A