Sensidyne Gilian 801961 Low Flow Adapter for 3500 & 5000 Air Sampling Pumps
Representative image only.
Sensidyne Gilian 801961 Low Flow Adapter for 3500 & 5000 Air Sampling Pumps
Sensidyne | Mfr: 801961| Item: EW-86276-73
Optimize performance of your air/gas analyzer
$251.50USD / Each
$251.50USD / Each
Product Description
Key Features
  • Airflow indicator
  • Smoke tubes
  • Low flow adapter
  • Achieve a flow rate of 20 to 750 cc/min
  • 3 month warranty
More About this Item
Low flow adapter, air indicator kit, and other accessories allow you to get the most out of your sampling and analysis system.
This adapter allows for lower sampling flow rates for the Gillian 3500 and 5000 Sampling Pumps.
3 Month Warranty
Specifications
  • Compatible WithGilian Models 3500 and 5000
  • DescriptionLow Flow Adapter for 3500 & 5000 Air Sampling Pumps, 20 to 750 cc/min