SmartTool Angle Sensor Module, Digital Protractor/Level
Representative image only.
SmartTool Angle Sensor Module, Digital Protractor/Level
| Item: EW-09918-06
$204.75USD / Each
for an additional
$110.50USD / Each
for an additional
$196.00USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$204.75USD / Each
Buy It Together
Current Item
SmartTool Angle Sensor Module, Digital Protractor/Level
SmartTool Angle Sensor Module, Digital Protractor/Level
0991806
$204.75USD / Each
+ =
Total Price 315.25 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Digital Level
This unit is designed to be job site tough. The box frame is extruded from high strength aluminum. The two bubble vials are machined from solid acrylic blocks and are virtually indestructible. The unit's Sensor Module is encased in a high impact resistant ABS polymer case, sealed against dirt and water penetration.
The Digital Level can be recalibrated in seconds with the touch of a button.
Pitch
Gives roof pitch readings -- in inches of rise per foot of run — right on the display. No two-handed measurements, no conversion calculations and no mistakes.
Reads In Degrees
Set saw blade angles to a fraction of a degree accuracy with SmartTool™. Easily duplicates a stair angle for setting hand rails
Listen & Level Audio Feature
The beeper sounds when SmartTool™ is level and/or plumb.
% Slope
Read angles in % slope to meet requirements for sewer connections, ADA ramps and grading. A
Available in 24" Length
Can be purchased as a complete system in 24-inch rail lengths. The 6.64-inch module can be used as a torpedo level for small tasks, or locked into any American brand carpentry and construction series levels.
1 Year Warranty
Specifications
  • DescriptionAngle Sensor Module, Digital Protractor/Level