Spectrum Chemical Barium Hydroxide, Octahydrate, Reagent ACS Grade; 500 g

Representative image only.

Spectrum Chemical Barium Hydroxide, Octahydrate, Reagent ACS Grade; 500 g

Spectrum Chemical | Mfr: B1040-500GM| Item: EW-80024-46

Quality and delivery you can count on every time!

$290.25USD / Each

$290.25USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaBa(OH)2•8H2O
  • Linear FormulaBa(OH)2•8H2O
  • ClassUN3262, Corrosive solid, basic, inorganic, n.o.s. (barium hydroxide), 8, PG III
  • Weight (g)500
  • Formula Weight315.46
  • PackagingPoly Bottle
  • CAS Number12230-71-6
  • UN NumberUN3262
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionBarium Hydroxide, Octahydrate, Reagent ACS Grade; 500 g
Documents & Certificates
Reviews
Q&A