Spectrum Chemical Dimethyl Sulfoxide, USP Grade; 100 mL

Representative image only.

Spectrum Chemical Dimethyl Sulfoxide, USP Grade; 100 mL

Spectrum Chemical | Mfr: D1258-100ML| Item: EW-80068-61

Quality and delivery you can count on every time!

$521.00USD / Each

$521.00USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeUSP
  • Empirical FormulaC2H6OS
  • Linear FormulaC2H6OS
  • SynonymsDMSO | Methyl Sulfoxide
  • Volume (mL)100
  • Formula Weight78.13
  • PackagingAmber Glass
  • Storage SymbolLIGHT SENSITIVE, HYGROSCOPIC: Keep tightly closed in light-resistant containers.
  • CAS Number67-68-5
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionDimethyl Sulfoxide, USP Grade; 100 mL
Documents & Certificates
Reviews
Q&A