Spectrum Chemical Hydrochloric Acid, Reagent, ACS; 4 L Plastic Bottle

Representative image only.

Spectrum Chemical Hydrochloric Acid, Reagent, ACS; 4 L Plastic Bottle

Spectrum Chemical | Mfr: H1035-4LTPL| Item: EW-10001-75

Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

$174.50USD / Each

$174.50USD / Each
Product Description

Key Features

  • Quality and delivery you can count on every time!

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • FormLiquid
  • Volume (mL)4000
  • CAS Number7647-01-0
  • DescriptionSpectrum Chemical Hydrochloric Acid, Reagent, ACS; 4 L Plastic Bottle
Documents & Certificates
Reviews
Q&A