Spectrum Chemical Hydrogen Peroxide, 30% Solution Reagent ACS Grade; 500 mL

Representative image only.

Spectrum Chemical Hydrogen Peroxide, 30% Solution Reagent ACS Grade; 500 mL

Spectrum Chemical | Mfr: H1065-500MLPL| Item: EW-80100-22

Quality and delivery you can count on every time!

$286.75USD / Each

$286.75USD / Each

Buy It Together

Current Item

Spectrum Chemical Hydrogen Peroxide, 30% Solution Reagent ACS Grade; 500 mL
Spectrum Chemical Hydrogen Peroxide, 30% Solution Reagent ACS Grade; 500 mL
8010022

$286.75USD / Each
+ =
Total Price 349.45 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaH2O2
  • Linear FormulaH2O2
  • ClassUN2014, Hydrogen peroxide, aqueous solution, 5.1 (8), PG II
  • Volume (mL)500
  • Formula Weight34.01
  • PackagingPoly Bottle
  • Storage SymbolREFRIGERATE: Keep tightly closed.
  • UN NumberUN2014
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionHydrogen Peroxide, 30% Solution Reagent ACS Grade; 500 mL
Reviews
Q&A