Spectrum Chemical Methylene Chloride, Technical Grade; 4 L

Representative image only.

Spectrum Chemical Methylene Chloride, Technical Grade; 4 L

Spectrum Chemical | Mfr: M1252-4LTGL| Item: EW-80127-54

Quality and delivery you can count on every time!

$292.25USD / Each

$292.25USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Empirical FormulaCH2Cl2
  • Linear FormulaCH2Cl2
  • ClassUN1593, Dichloromethane, 6.1, PG III
  • SynonymsDichloromethane
  • Volume (mL)4000
  • Formula Weight84.93
  • PackagingAmber Glass
  • UN NumberUN1593
  • CAS Number75-09-2
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionMethylene Chloride, Technical Grade; 4 L
Documents & Certificates
Reviews
Q&A