Spectrum Chemical n-Decane; 100 mL

Representative image only.

Spectrum Chemical n-Decane; 100 mL

Spectrum Chemical | Mfr: D2098-100ML| Item: EW-80071-72

Quality and delivery you can count on every time!

$108.75USD / Each

$108.75USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity Grade
  • Empirical FormulaC10H22
  • Linear FormulaC10H22
  • ClassUN2247, n-Decane, 3, PG III
  • Volume (mL)100
  • Formula Weight142.28
  • PackagingAmber Glass
  • UN NumberUN2247
  • CAS Number124-18-5
  • Shelf Life (days)730
  • Shelf Life (months)24
  • Descriptionn-Decane; 100 mL
Documents & Certificates
Reviews
Q&A