Spectrum Chemical Perchloric Acid, 60%, Reagent ACS Grade; 500 mL

Representative image only.

Spectrum Chemical Perchloric Acid, 60%, Reagent ACS Grade; 500 mL

Spectrum Chemical | Mfr: P1030-500MLGL| Item: EW-80151-61

Quality and delivery you can count on every time!

$381.00USD / Each

$381.00USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaHClO4
  • Linear FormulaHClO4
  • ClassUN1873, Perchloric acid, 5.1 (8), PG I
  • Volume (mL)500
  • Formula Weight100.46
  • PackagingAmber Glass
  • Storage SymbolHYGROSCOPIC: Keep tightly closed
  • CAS Number7601-90-3
  • UN NumberUN1873
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionPerchloric Acid, 60%, Reagent ACS Grade; 500 mL
Documents & Certificates
Reviews
Q&A