Spectrum Chemical Quinhydrone; 25 g

Representative image only.

Spectrum Chemical Quinhydrone; 25 g

Spectrum Chemical | Mfr: Q2010-25GM| Item: EW-80181-65

Quality and delivery you can count on every time!

$196.00USD / Each

$196.00USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity Grade
  • Empirical FormulaC6H4O2•C6H4(OH)2
  • Linear FormulaC6H4O2•C6H4(OH)2
  • ClassUN2811, Toxic solid, organic, n.o.s. (quinhydrone), 6.1, PG III
  • Weight (g)25
  • Formula Weight218.21
  • PackagingAmber Glass
  • Storage SymbolLIGHT SENSITIVE: Keep tightly closed in light-resistant containers.
  • CAS Number106-34-3
  • UN NumberUN2811
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionQuinhydrone; 25 g
Documents & Certificates
Reviews
Q&A