Spectrum Chemical Sodium Silicofluoride, Reagent Grade; 25 g

Representative image only.

Spectrum Chemical Sodium Silicofluoride, Reagent Grade; 25 g

Spectrum Chemical | Mfr: S1440-25GM| Item: EW-80194-42

Quality and delivery you can count on every time!

$94.10USD / Each

$94.10USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeReagent
  • Empirical FormulaNa2SiF6
  • Linear FormulaNa2SiF6
  • ClassUN2674, Sodium fluorosilicate, 6.1, PG III
  • ChemistrySolutions
  • SynonymsSodium Fluorosilicate
  • Weight (g)25
  • Formula Weight188.06
  • PackagingPoly Bottle
  • CAS Number16893-85-9
  • UN NumberUN2674
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionSodium Silicofluoride, Reagent Grade; 25 g
Documents & Certificates
Reviews
Q&A