Spectrum Chemical Sulfuric Acid, Reagent, ACS; 500 mL Glass Bottle

Representative image only.

Spectrum Chemical Sulfuric Acid, Reagent, ACS; 500 mL Glass Bottle

Spectrum Chemical | Mfr: S1730-500MLGL| Item: EW-10000-94

Quality and delivery you can count on every time!

$102.50USD / Each

$102.50USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • FormLiquid
  • Volume (mL)500
  • CAS Number7664-93-9
  • DescriptionSpectrum Chemical Sulfuric Acid, Reagent, ACS; 500 mL Glass Bottle
Documents & Certificates
Reviews
Q&A