Spectrum Chemical Tetrahydrofuran Stabilized, Technical Grade; 4 L

Representative image only.

Spectrum Chemical Tetrahydrofuran Stabilized, Technical Grade; 4 L

Spectrum Chemical | Mfr: T1226-4LTGL| Item: EW-80220-26

Quality and delivery you can count on every time!

$494.50USD / Each

$494.50USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Empirical FormulaC4H8O
  • Linear FormulaC4H8O
  • ClassUN2056, Tetrahydrofuran, 3, PG II
  • SynonymsTHF
  • Volume (mL)4000
  • Formula Weight72.11
  • PackagingAmber Glass
  • UN NumberUN2056
  • CAS Number109-99-9
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionTetrahydrofuran Stabilized, Technical Grade; 4 L
Reviews
Q&A