Spectrum Chemical Triisopropanolamine; 1 kg

Representative image only.

Spectrum Chemical Triisopropanolamine; 1 kg

Spectrum Chemical | Mfr: T2120-1KG| Item: EW-80226-54

Quality and delivery you can count on every time!

$158.25USD / Each

$158.25USD / Each
Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity Grade
  • Empirical FormulaC9H21NO3
  • Linear FormulaC9H21NO3
  • Synonyms1,1',1"-Nitrilotripropan-2-ol
  • Weight (g)1000
  • Formula Weight191.27
  • PackagingAmber Glass
  • Storage SymbolHYGROSCOPIC: Keep tightly closed.
  • CAS Number122-20-3
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionTriisopropanolamine; 1 kg
Reviews
Q&A