Spex Cupriethylenediamine, 1.0 M, Amber Glass Bottle; 4 L

Representative image only.

Spex Cupriethylenediamine, 1.0 M, Amber Glass Bottle; 4 L

Spex | Item: EW-84410-16

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$433.00USD / Each

$433.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 4 L amber glass bottle

More About this Item

Density: 1.1
Composition: water 84.10%, cupriethylenediamine 15.90%
Solubility: miscible
Color: dark blue liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Concentration1 M
  • ChemistrySolvents
  • Element/IonCupriethylenediamine
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationHazardous; CUPRIETHYLENEDIAMINE SOLUTION; Class 8 (6.1), Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosive
  • CAS Number13426-91-0
  • UN NumberUN1761
  • Shelf Life (months)12
  • DescriptionCupriethylenediamine, 1.0 M, Amber Glass Bottle; 4 L
Documents & Certificates
Reviews
Q&A