Spex Hydrochloric Acid, ACS Grade, 37% w/w, Glass Bottle; 2.5 L

Representative image only.

Spex Hydrochloric Acid, ACS Grade, 37% w/w, Glass Bottle; 2.5 L

Spex | Item: EW-84410-28

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$121.00USD / Each

$121.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 2.5 L safety-coated clear glass bottle

More About this Item

Solubility: miscible
Color: colorless liquid

Density: 1.18
Molecular weight: 36.46g/mol
Composition: hydrochloric acid 37%, water 63%
pH range: < 1

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Purity GradeACS
  • Concentration37%
  • ChemistrySolvents
  • Element/IonHydrochloric Acid
  • FormLiquid
  • Volume (mL)2500
  • PackagingClear glass bottle, safety coated
  • DOT InformationHazardous; Hydrochloric Acid; Class 8, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN1789
  • CAS Number7647-01-0
  • Shelf Life (months)36
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)-28
  • Melting Point F (° F)-18.4
  • DescriptionHydrochloric Acid, ACS Grade, 37% w/w, Glass Bottle; 2.5 L
Documents & Certificates
Reviews
Q&A