Spex Isopropanol, 99%, Anhydrous, Purified; 20 L

Representative image only.

Spex Isopropanol, 99%, Anhydrous, Purified; 20 L

Spex | Item: EW-84410-39

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$709.00USD / Each

$709.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 20 L round metal can

More About this Item

Assay percentage: 0.99
Flash point: 12 °C
Density: 0.78
Molecular weight: 60.1 g/mol
Chemical formula: CH3CHOHCH3
Composition: isopropyl Alcohol 100%
Solubility: miscible
Refractive Index: 1.376 to 1.378nD
Color: colorless liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Purity GradeAnhydrous, Purified
  • Linear FormulaCH3CHOHCH3
  • ChemistrySolvents
  • Element/IonIsopropanol
  • FormLiquid
  • Synonyms2-Propanol, dimethyl carbinol, IPA, isopropanol, rubbing alcohol
  • Volume (mL)20000
  • PackagingRound metal can
  • DOT InformationHazardous; Isopropanol; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant
  • CAS Number67-63-0
  • UN NumberUN1219
  • Shelf Life (months)240
  • Boiling Point C (° C)82.5
  • Boiling Point F (° F)180.5
  • Melting Point C (° C)-90
  • Melting Point F (° F)-130
  • DescriptionIsopropanol, 99%, Anhydrous, Purified; 20 L
Documents & Certificates
Reviews
Q&A